Learn More
Abnova™ LY6D 293T Cell Transient Overexpression Lysate (Denatured) (T04)
Human overexpression lysate
Manufacturer:  Abnova™ H00008581T04
Catalog No. 89-951-270
Add to cart
Description
Description
Specifications
Specifications
293T
BC031330
pCMV-LY6D full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
LY6D 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 11.99kDa
Transient overexpression cell lysate was tested with anti-LY6D antibody (Mfr. No. H00008581-B03) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human