Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LY6G6F Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107164
Description
Sequence homology: Cow: 86%; Dog: 85%; Guinea Pig: 77%; Horse: 92%; Human: 100%; Rabbit: 100%.
May play a role in the downstream signal transduction pathways involving GRB2 and GRB7.
Specifications
LY6G6F | |
Polyclonal | |
Unconjugated | |
LY6G6F | |
C6orf21; G6F; LY6G6D; LY6G6F; Lymphocyte antigen 6 complex locus protein G6f; lymphocyte antigen 6 complex, locus G6D; lymphocyte antigen 6 complex, locus G6F; NG32 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
259215 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
Q5SQ64 | |
LY6G6F | |
Synthetic peptide directed towards the middle region of human C6orf21 (aa 144-194). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction