Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

LY6G6F Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548799

Catalog No. PA548799



Description

Description

May play a role in the downstream signal transduction pathways involving GRB2 and GRB7.
Specifications

Specifications

LY6G6F
Polyclonal
Unconjugated
LY6G6F
Liquid
LY6G6F
KLH conjugated synthetic peptide between 56-84 amino acids from the N-terminal region of human LY6G6F.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q5SQ64
Lymphocyte antigen 6 complex locus protein G6f, LY6G6F, C6orf21, G6F, LY6G6D, NG32
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
259215
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit