Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LY6K Mouse anti-Human, Clone: CL2433, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524638

Catalog No. PIMA524638


Add to cart

Description

Description

Immunogen sequence: TDEGDNRVWC HVCERENTFE CQNPRRCKWT EPYCVIAAVK IFPRFFMVAK QCSAGCAAME RPKPEEKRFL LEEPMPFFYL KCCKIRYCNL EGPPINSSVF KEYAG Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 36%. Binds to an epitope located within the peptide sequence LEEPMPFFYLKCCKI as determined by overlapping synthetic peptides.

Granulocytes are a category of white blood cells characterised by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.
Specifications

Specifications

LY6K
Monoclonal
1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q17RY6
CO16, CT97, cancer/testis antigen 97, CO16, CT97, FLJ35226, HSJ001348, Ly-6K, ly-6K, lymphocyte antigen 6 complex, locus K, lymphocyte antigen 6K
Mouse
IgG1
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
54742
Immunohistochemistry, Western Blot
CL2433
Unconjugated
LY6K
Liquid
LY6K
Recombit protein corresponding to Human LY6K
Protein A
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Research Use Only