Learn More
CD229 Mouse anti-Human, PE, Clone: HLy9.25, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen A15791
Catalog No. 50-113-7108
Add to cart
Description
Description
CD229 (Ly9) is a cell surface receptor of the CD150 family, which includes also e.g. CD48 and CD224. Receptors of this family regulate cytokine production and cytotoxicity of lymphocytes and NK cells. High levels of CD229 are found on T and B cells, where its expression increases during their maturation. It is absent on granulocytes, bone marrow-derived dendritic cells, platelets and erythrocytes. CD229 has been also reported on mouse monocytes and NK cells. CD229 interacts homophilically through its N-terminal domain and localizes to the contact site between T cells and antigen presenting B cells during antigen-dependent immune synapse formation.
Specifications
Specifications
CD229
Monoclonal
PE
LY9
Liquid
LY9
Human CD229
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
HLy9.25
PBS with 0.2% BSA and 15mM sodium azide
Q9HBG7
RP11-312J18.1, CD229, SLAMF3, hly9, mLY9, T-lymphocyte surface antigen Ly-9, cell surface molecule Ly-9, cell-surface molecule Ly-9
Mouse
IgG1
100 tests
4° C
Primary
4063