Learn More
Aviva Systems Biology Corp LYAR Antibody - middle region (ARP57042_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP57042_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LYAR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LYAR. Alternate names: FLJ20425, ZLYARApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-62067