Learn More
Lyle and Louise: Stained with Suspicion™ Blood Detection and Evidence Processing
SCI_ED
Introduces students to presumptive blood tests and evidence processing.
Supplier:  Crosscutting Concepts LLC VXHLLBDDRF
 View more versions of this product
Catalog No. S02173R
Add to Cart
Includes: Six bottles of hydrogen peroxide, six bottles of phenolphthalein solution, six sets of cotton swabs, six sets of positive controls and presumptive test cards, six carpet scraps from the crime scene, fully illustrated teacher's manual with technique background
Requires : Lab gloves, distilled water, rulers for each group, camera (optional), tweezers (optional)
Description
Description
Helps determine if a substance at the crime scene is blood and teaches forensic techniques involved in evidence processing.
  • Analyze different items to find traces of blood
  • Process the piece of evidence as processed at the actual crime scene
Order Info
For up to 30 students working in six groups. Kit is geared toward high school and undergraduate courses and matched to National Science Education Standards.
Specifications
Specifications
Mystery of Lyle and Louise: Stained with Suspicion™ Blood Detection and Evidence Processing Forensic Science Refill Kit
For introduces students to presumptive blood tests and evidence processing
Lab gloves, distilled water, rulers for each group, camera, tweezers
9 to Undergraduate
Six bottles of Hydrogen Peroxide, six bottles of Phenolphthalein solution, six sets of Cotton swabs, six sets of positive controls and presumptive test cards, six carpet scraps from the crime scene, fully illustrated teacher's manual with technique background
30 Students