Learn More
Lonza™ BioWhittaker™ Lymphocyte Separation Medium
Intended for laboratory/manufacturing use.   BioWhittaker™ Lymphocyte Separation Medium is commonly used to isolate lymphocytes from human blood.
Manufacturer:  Lonza™ 17829E/24
 View more versions of this product
Catalog No. BW17829E24
Add to cart
This item is not returnable. View return policy
Description
Description
  • Mixture of Ficoll™ and sodium diatrizoate (Hypaque)
  • Density adjusted to 1.077 g/ml
  • Sterile filtered
  • Intended for laboratory/manufacturing use
  • Not for in vitro diagnostic use
Specifications
Specifications
Lymphocyte separation medium
15° to 30°C
Sterile
24 x 100mL