Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lymphotoxin-alpha/TNF-beta Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0575-7U9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212441AF532

Catalog No. NP212441A32


Add to cart

Description

Description

Lymphotoxin-alpha/TNF-beta Monoclonal antibody specifically detects Lymphotoxin-alpha/TNF-beta in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Lymphotoxin-alpha/TNF-beta
MM0575-7U9
Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
LTA
Human
Protein G purified
Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity
Primary
4049.0
Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
LT, LT-alpha, lymphotoxin alpha (TNF superfamily, member 1), TNFBlymphotoxin-alpha, TNFSF1TNF-beta, tumor necrosis factor beta, Tumor necrosis factor ligand superfamily member 1
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit