Learn More
Lymphotoxin-alpha/TNF-beta Mouse anti-Human, Alexa Fluor™ 750, Clone: MM0575-7U9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212441AF750
Catalog No. NP212441A70
Add to cart
Description
Description
Lymphotoxin-alpha/TNF-beta Monoclonal antibody specifically detects Lymphotoxin-alpha/TNF-beta in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
Lymphotoxin-alpha/TNF-beta
MM0575-7U9
Immunohistochemistry, Immunohistochemistry-Paraffin
LT, LT-alpha, lymphotoxin alpha (TNF superfamily, member 1), TNFBlymphotoxin-alpha, TNFSF1TNF-beta, tumor necrosis factor beta, Tumor necrosis factor ligand superfamily member 1
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
LTA
Human
0.1 mL
Primary
4049