Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lymphotoxin-alpha/TNF-beta, Rabbit anti-Human, Clone: 3F12.2D3, Novus Biologicals™

Rabbit Monoclonal Antibody

$156.50 - $419.00

Specifications

Antigen Lymphotoxin-alpha/TNF-beta
Clone 3F12.2D3
Applications Flow Cytometry, ELISA, Neutralization
Host Species Rabbit
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB016183
View Documents
Novus Biologicals
NBP2811050.025MG
0.025 mg
Each for $156.50
Add to cart
 
NB016184
View Documents
Novus Biologicals
NBP2811050.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

Lymphotoxin-alpha/TNF-beta Monoclonal antibody specifically detects Lymphotoxin-alpha/TNF-beta in Human Samples. It is validated for Flow Cytometry, ELISA, Surface Plasmon Resonance, Neutralization.
Specifications

Specifications

Lymphotoxin-alpha/TNF-beta
Flow Cytometry, ELISA, Neutralization
Human
Primary
3F12.2D3
Rabbit
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit