Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lymphotoxin beta R/TNFRSF3 Antibody (MM0461-3T24) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2119470.025MG
Description
Lymphotoxin beta R/TNFRSF3 Monoclonal specifically detects Lymphotoxin beta R/TNFRSF3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
Lymphotoxin beta R/TNFRSF3 | |
Monoclonal | |
LYOPH | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, CyTOF-ready | |
P36941 | |
LTBR | |
Human recombinant protein of Lymphotoxin-beta Receptor extracellular domain. | |
0.025 mg | |
Apoptosis, Cytokine Research | |
4055 | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
MM0461-3T24 | |
Unconjugated | |
A 0.2 um filtered solution in PBS with No Preservative | |
CD18, D12S370, ltbetar, LT-BETA-R, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction