Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lymphotoxin beta R/TNFRSF3 Rat anti-Mouse, Allophycocyanin/Cy7, Clone: 3C8, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP143308APCCY7
Description
Lymphotoxin beta R/TNFRSF3 Monoclonal antibody specifically detects Lymphotoxin beta R/TNFRSF3 in Mouse samples. It is validated for Flow Cytometry.Specifications
Lymphotoxin beta R/TNFRSF3 | |
3C8 | |
Flow Cytometry | |
Conjugated, Purified | |
LTBR | |
The immunogen for this antibody was Lymphotoxinbeta Receptor. | |
Protein A or G purified | |
Apoptosis, Cytokine Research | |
Primary | |
4055.0 |
Flow Cytometry | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
CD18, D12S370, ltbetar, LT-BETA-R, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III | |
Rat | |
IgG1 Kappa | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title