Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lymphotoxin beta R/TNFRSF3 Rat anti-Mouse, Clone: RM0341-11N1, Novus Biologicals™
Rat Monoclonal Antibody
$177.16 - $493.00
Specifications
Antigen | Lymphotoxin beta R/TNFRSF3 |
---|---|
Clone | RM0341-11N1 |
Concentration | LYOPH |
Applications | Western Blot, Flow Cytometry, CyTOF |
Classification | Monoclonal |
Description
Lymphotoxin beta R/TNFRSF3 Monoclonal specifically detects Lymphotoxin beta R/TNFRSF3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
Lymphotoxin beta R/TNFRSF3 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
CD18, D12S370, ltbetar, LT-BETA-R, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III | |
LTBR | |
Primary |
RM0341-11N1 | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
Rat | |
Apoptosis, Cytokine Research | |
P50284 | |
4055 | |
IgG2 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title