Learn More
Lymphotoxin beta R/TNFRSF3 Rat anti-Mouse, DyLight 650, Clone: 3C8, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143308C
Catalog No. NBP143308C
Add to cart
Description
Description
Lymphotoxin beta R/TNFRSF3 Monoclonal antibody specifically detects Antigen in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
Lymphotoxin beta R/TNFRSF3
3C8
Flow Cytometry
Purified
LTBR
The immunogen for this antibody was Lymphotoxinbeta Receptor.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Mouse
Flow Cytometry
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
CD18, D12S370, ltbetar, LT-BETA-R, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III
Rat
IgG1 Kappa
0.1 ml
Apoptosis, Cytokine Research
Primary
4055