Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYN Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 181351AP

Catalog No. 50-173-2005


Add to cart

Description

Description

Prohibitin is an evolutionarily conserved gene that is ubiquitously expressed. It is thought to be a negative regulator of cell proliferation and may be a tumor suppressor. Mutations in PHB have been linked to sporadic breast cancer. Prohibitin is expressed as two transcripts with varying lengths of 3 ft. untranslated region. The longer transcript is present at higher levels in proliferating tissues and cells, suggesting that this longer 3 ft. untranslated region may function as a trans-acting regulatory RNA.
Specifications

Specifications

LYN
0.39 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P07948
JTK8, LYN, Tyrosine protein kinase Lyn
Rabbit
IgG
150 μL
-20°C
Primary
4067
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
LYN
Liquid
LYN
LYN Fusion Protein Ag12723
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit