Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lyon™ Plastic Storage Bins

Features one full-width shelf

$5.03 - $36.30

Specifications

Material Plastic
Product Type Storage Bin
For Use With (Equipment) Wall cabinet
Includes One full-width shelf, three adjustable half-shelves, 50 small and 14 large Plastic parts bins
View More Specs

Products 2
Catalog Number Mfr. No. Height (English) Length (English) Width (English) Size Price Quantity & Availability  
Catalog Number Mfr. No. Height (English) Length (English) Width (English) Size Price Quantity & Availability  
19-813-252
View Documents
Lyon™
78222
3 in. 5.37 in. 4.12 in. S Each for $5.03
Add to cart
 
19-813-253
View Documents
Lyon™
78228
7 in. 14.75 in. 8.12 in. L Each for $36.30
Add to cart
 
Description

Description

  • Three half-width shelves adjustable on 3 in. centers
  • 50 small, 10.5 x 13.67 x 7.62cm (4.13 in. x 5.38 in. x 3 in.), plastic parts bins on door
  • 14 large, 21 x 37.47 x 17.78cm (8.25 in. x 14.75 in. x 7 in.), plastic parts bins inside cabinet

Specifications

Specifications

Plastic
Storage Bin
Wall cabinet
One full-width shelf, three adjustable half-shelves, 50 small and 14 large Plastic parts bins
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit