Learn More
Innovating Science™ LyoQuick Freeze Dried Bacteria Cultures

Easy to use lyophilized bacterial cultures for use in classroom for educational purposes. Innovating Science™ LyoQuick Freeze Dried Bacteria Cultures, eliminates several steps required for rehydration of freeze-dried cultures, thus reducing the risk of contamination.
Specifications
Color | White |
---|---|
Grade | 7 to 14 |
Age | 13 to 20 |
Includes | LyoQuick Freeze Dried Bacteria |
Quantity | 1 |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S29044
![]() |
Innovating Science™
IS3602 |
E. coli | Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. | N/A | |||||
S29047
![]() |
Innovating Science™
IS3606 |
Staphylococcus epidermidis | Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. | N/A | |||||
S37735
![]() |
Innovating Science™
IS3609 |
E. aerogenes | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. | N/A | |||||
Description
- Each tube contains two ampules, one containing freeze-dried bacteria and one self-contained sterile rehydration broth
- To use, pinch tube to break internal ampules and let freeze-dried organisms rehydrate, then dispense rehydrated organisms into broth or onto agar through dropper tip
- Available for eight different types of bacteria
Specifications
White | |
13 to 20 | |
1 |
7 to 14 | |
LyoQuick Freeze Dried Bacteria | |
4 x 0.5 x 0.5 in. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.