Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BUCHI Lyovapor™ L-200 Freeze Dryer

The compact Lyovapor™ L-200 Classic offers high-quality Freeze Drying with a high level of automation.
Supplier: BUCHI 11L200IN0102000002
Description
- A 6 kg condenser capacity allows for maximum sample throughput
- Integrating Infinite-Control™ with your freeze dryer gives you access to all process parameters and keeps you informed by push notifications anywhere, anytime
- Fully Automated
- Method creation and reporting
- Real-time pressure and temperature recording
- Stoppering available under inert atmospheric conditions
- Small footprint
- Touch-screen control panel
Certifications
CE Certified
Specifications
6 kg (Condensor) | |
Stainless Steel | |
Manifold Rack 12 | |
510 mm | |
349.7 lb. |
-55°C to +25°C | |
Freeze Drying | |
585 x 460 x 510 mm | |
208/230 V | |
159 kg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title