Learn More
BUCHI Lyovapor™ L-200 Classic Freeze Dryer: PROMO
The compact Lyovapor™ L-200 Classic offers high-quality Freeze Drying with a high level of automation.
Manufacturer:  BUCHI 11L200IN1421000102PM
 View more versions of this product
Catalog No. 05-403-003PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Modular platform for future upgrades
  • Small footprint
  • Touch-screen control panel
  • Status messages on process intervals
  • Cost Effective
Certifications
CE Certified
Specifications
Specifications
Up to 55°C
50/60 Hz
74 kg
23 in.
CE Certified
208/230 V
163 lb.
6 L
20 in.
Pfieffer Duo 6 Vacuum Pump, Tray 4-position and Manifold 12-position