Learn More
BUCHI Lyovapor™ L-200 Classic Freeze Dryer PROMO
The compact Lyovapor™ L-200 Classic offers high-quality Freeze Drying with a high level of automation.
Manufacturer:  BUCHI 11L200IN1421000102PR
 View more versions of this product
Catalog No. 05-403-003PR
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Modular platform for future upgrades
  • Small footprint
  • Touch-screen control panel
  • Status messages on process intervals
  • Cost Effective
Specifications
Specifications
CE Certified
Pfieffer Duo 6 Vacuum Pump, Tray 4-position and Manifold 12-position
23 in.
163 lb.
Upto 55°C
50 to 60 Hz
6L
20 in.
74kg
208 to 230V