Learn More
BUCHI Lyovapor™ L-200 Pro Control Freeze Dryer: PROMO
The compact Lyovapor™ L-200 Pro Control offers high-quality Freeze Drying with complete automation.
Manufacturer:  BUCHI 11L200PH2422111002PM
 View more versions of this product
Catalog No. 05-403-005PM
Add to cart
Description
Description
  • A 6L condenser capacity allows for maximum sample throughput
  • Integrating Infinite-Control™ with your freeze dryer gives you access to all process parameters and keeps you informed by push notifications anywhere, anytime
  • Fully Automated
  • Method creation and reporting
  • Real-time pressure and temperature recording
  • Stoppering available under inert atmospheric conditions
  • Small footprint
  • Touch-screen control panel
Certifications
CE Certified
Specifications
Specifications
50 to 60 Hz
6L
20 in.
74kg
208 to 230V
Pfieffer Duo 6 Vacuum Pump, Heated shelves 4-position and Manifold 12-position
23 in.
163 lb.
−55°C