Learn More
BUCHI Lyovapor™ L-300 Continuous Freeze Dryer
The Continuous L-300 Lyovapor™ is a unique freeze dryer, providing unlimited freeze drying for the first time with BUCHI's Infinite-Technology™. Dual condensers allow for continuous sublimation of both water and organic based solvents.
Manufacturer:  BUCHI 11L300IN0305002004
 View more versions of this product
Catalog No. 05-403-008
Add to cart
Description
Description
  • Infinite-Technology™, allows for continuous freeze drying due to dual condensers
  • Lowest Condensor Temperature: −105°C
  • Automated cleaning
  • Sublimation of both water and organic-based solvents
  • Advanced software for creating and managing methods, data record and reporting
Certifications
CE Certified
Specifications
Specifications
Manifold Rack 36
250 kg
551 lb.
Freeze Drying
Stainless Steel
208/230 V
-105°C to 25°C
12 kg (Condensor)
39 in.
35 in.
CSA