Learn More
BUCHI Lyovapor™ L-300 Continuous Freeze Dryer PROMO
The Continuous L-300 Lyovapor™ is a unique freeze dryer, providing unlimited freeze drying for the first time with BUCHI's Infinite-Technology™. Dual condensers allow for continuous sublimation of both water and organic based solvents.
Manufacturer:  BUCHI 11L300IN0305002004PR
 View more versions of this product
Catalog No. 05-403-008PR
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Infinite-Technology™, allowing for continuous freeze drying due to dual condensers
  • −105°C lowest condenser temperature
  • Automated cleaning
  • Sublimiation of both water and organic-based solvents
  • Advanced software for creating and managing methods, data record and reporting
Specifications
Specifications
−105°C
208 to 230V
Pfieffer Duo 11 Vacuum Pump, Manifold 36-position
39 in.
551 lb.
3-phase
CE Certified
unlimited
35 in.
250kg