Learn More
BUCHI Lyovapor™ L-300 Continuous Pro Modular Freeze Dryer
The Continuous L-300 Lyovapor™ Pro Modular is a unique freeze dryer, providing unlimited freeze drying for the first time with BUCHI's Infinite-Technology™. Dual condensers allow for continuous sublimation of both water and organic based solvents.
Manufacturer:  BUCHI 11L300PH2618112004
 View more versions of this product
Catalog No. 05-403-010
Add to cart
Description
Description
  • Infinite-Technology™, allowing for continuous freeze drying due to dual condensers
  • −105°C lowest condenser temperature
  • Automated cleaning
  • Sublimiation of both water and organic-based solvents
  • Advanced software for creating and managing methods, data record and reporting
Certifications
CE Certified
Specifications
Specifications
12 kg (Condensor)
35 in.
39 in.
250 kg
Stainless Steel
208/230 V
Heatable Shelves 6, IVC + DC Capacitive and Pirani Vacuum Sensor, Temperature Sensor, Software, Edwards nXDS10iC Vacuum Pump
551 lb.
-105°C to 25°C
Freeze Drying
CSA