Learn More
BUCHI Lyovapor™ L-300 Continuous Pro Modular Freeze Dryer: PROMO
The Continuous L-300 Lyovapor™ Pro Modular is a unique freeze dryer, providing unlimited freeze drying for the first time with BUCHI's Infinite-Technology™. Dual condensers allow for continuous sublimation of both water and organic based solvents.
Manufacturer:  BUCHI 11L300PH2618112004PM
 View more versions of this product
Catalog No. 05-403-010PM
Add to cart
Description
Description
  • Infinite-Technology™, allowing for continuous freeze drying due to dual condensers
  • -105°C lowest condenser temperature
  • Automated cleaning
  • Sublimiation of both water and organic-based solvents
  • Advanced software for creating and managing methods, data record and reporting
Certifications
CE Certified
Specifications
Specifications
CE Certified
unlimited
35 in.
250kg
3-phase
Pfieffer Duo 11 Vacuum Pump, Heatable shelves 6-position
39 in.
551 lb.
−105°C
208 to 230V