Learn More
LYPD6B Mouse anti-Human, Mouse, Clone: 634CT7.10.1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524725
Catalog No. PIMA524725
Add to cart
Description
Description
Believed to act as a modulator of nicotinic acetylcholine receptors (nAChRs) activity. In vitro acts on nAChRs in a subtype- and stoichiometry-dependent manner. Modulates specifally alpha-3&No.8323;:beta-4&No.8322; nAChRs by enhancing the sensitivity to ACh, decreasing ACh-induced maximal current response and increasing the rate of desensitization to ACh; has no effect on alpha-7 homomeric nAChRs; modulates alpha-3&No.8322;:alpha-5:beta-4&No.8322; nAChRs in the context of CHRNA5/alpha-5 variant Asn-398 but not its wild-type sequence.
Specifications
Specifications
LYPD6B
Monoclonal
Unconjugated
LYPD6B
Liquid
LYPD6B
KLH conjugated synthetic peptide between 79-108 amino acids from the Central region of human LYPD6B
100 μL
-20°C
Primary
130576, 71897
Western Blot
634CT7.10.1
ascites with 0.09% sodium azide
Q8NI32, Q9D7F2
Ly6/PLAUR domain-containing protein 6B, LYPD6B
Mouse
IgG2b
RUO
Antibody
Monoclonal
Human, Mouse