Learn More
LYPD8 Mouse anti-Human, Alexa Fluor™ 350, Clone: 961703, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB9087U100UG
Catalog No. FAB9087U10
Add to cart
This item is not returnable. View return policy
Description
Description
LYPD8 Monoclonal antibody specifically detects LYPD8 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
LYPD8
961703
Flow Cytometry 0.25-1 μg/106 cells
Q6UX82
Mouse
IgG1
100 μg
Primary
646627
Human
Flow Cytometry
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Human embryonic kidney cell line HEK293-derived transfected with human LYPD8, Met1-Asn215, Accession # Q6UX82
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human LYPD8 in direct ELISAs. Stains human LYPD8 transfectants but not irrelevant transfectants in flow cytometry.