Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LYPLA1, Rabbit anti-Human,Mouse,Rat, Clone: JE47-60, Novus Biologicals
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP280466
Description
LYPLA1 Monoclonal antibody specifically detects LYPLA1 in Human, Mouse, Rat Samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
LYPLA1 | |
JE47-60 | |
Unconjugated | |
1X TBS(pH7.4), 1%BSA, 40%Glycerol | |
acyl-protein thioesterase-1, APT1, APT-1, EC 3.1.2, EC 3.1.2.-, hAPT1, LPL1acyl-protein thioesterase 1, LPL-I, Lysophospholipase 1, lysophospholipase ILYSOPLA, lysophospholipid-specific lysophospholipase, LysoPLA I | |
Rabbit | |
IgG | |
100 uL | |
Core ESC Like Genes, Stem Cell Markers | |
Primary | |
10434 |
Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
Purified | |
LYPLA1 | |
Recombinant protein within Lysophospholipase 1 aa 100-250. | |
Protein A purified | |
RUO | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human, Mouse, Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title