Learn More
LYPLA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 160551AP
Catalog No. 50-173-0681
Add to cart
Description
Description
Lysophospholipases are enzymes that act on biological membranes to regulate the multifunctional lysophospholipids. The protein encoded by this gene hydrolyzes lysophosphatidylcholine in both monomeric and micellar forms. The use of alternate polyadenylation sites has been found for this gene. There are alternatively spliced transcript variants described for this gene but the full length nature is not known yet.
Specifications
Specifications
LYPLA1
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75608, P70470, P97823
Acyl protein thioesterase 1, APT 1, APT1, hAPT1, LPL I, LPL1, LYPLA1, Lysophospholipase 1, lysophospholipase I, LYSOPLA, LysoPLA I
Rabbit
IgG
150 μL
-20°C
Primary
10434, 18777, 25514
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
LYPLA1
Liquid
Lypla1
LYPLA1 Fusion Protein Ag9022
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat