Learn More
Aviva Systems Biology Corp LYPLA2 Antibody - N-terminal region (ARP58638_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58638_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LYPLA2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LYPLA2. Alternate names: APT-2, DJ886K2.4ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62384