Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LYPLAL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545807
Description
Peptide sequence: EETNSMLKSL GVTTKFHSFP NVYHELSKTE LDILKLWILT KLPGEMEKQK Sequence homology: Dog: 82%; Guinea Pig: 100%; Horse: 85%; Human: 100%; Rabbit: 92%; Rat: 85%.
LYPLAL1 does not exhibit phospholipase nor triacylglycerol lipase activity. It is able to hydrolyze only short chain substrates due to its shallow active site.
Specifications
LYPLAL1 | |
Polyclonal | |
Unconjugated | |
Lyplal1 | |
BC027340; KIAA1238; LYPLAL1; lysophospholipase like 1; lysophospholipase-like 1; lysophospholipase-like protein 1; Q96AV0; Q96AVO | |
Rabbit | |
Affinity chromatography | |
RUO | |
127018 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5VWZ2 | |
Lyplal1 | |
Synthetic peptide directed towards the C-terminal of human LYPLAL1 (aa 188-237). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction