Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LYRM2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA520921

Catalog No. PIPA520921


Only null left
Add to Cart

Description

Description

A suggested positive control is A549 cell lysate. PA5-20921 can be used with blocking peptide PEP-1035.

The Lyr motif found in the LYR-motif containing protein family is similar to that found in the sacchromyces cerevisiae protein ISD11, an iron-sulfur protein in the mitochondria that is thought to play a role in iron homeostasis. No known function has been assigned to LYRM2, although LYRM1 is thought to be involved in preadipocyte progression and LYRM3 has been suggested to be a candidate gene for the branchio-oto-renal (BOR) syndrome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LYRM2
Polyclonal
Unconjugated
LYRM2
2610208E05Rik; DJ122O8.2; LYR motif containing 2; LYR motif-containing protein 2; Lyrm2; novel complex 1 (LYR family) protein
Rabbit
Antigen affinity chromatography
RUO
108755, 57226, 690354
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.02% sodium azide
B2GV91, Q8R033, Q9NU23
LYRM2
A 16 amino acid peptide near the carboxy terminus of human LYRM2.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.