Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A Mouse anti-Human, Clone: 3G10C2, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP261819
Description
Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A Monoclonal antibody specifically detects Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence.Specifications
Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A | |
3G10C2 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
EC 1.14.11, EC 1.14.11.-, JHDM2AJMJD1AJHMD2A, JmjC domain-containing histone demethylation protein 2A, JMJD1, jumonji C domain-containing histone demethylase 2A, jumonji domain containing 1, jumonji domain containing 1A, Jumonji domain-containing protein 1A, KIAA0742DKFZp686A24246, lysine (K)-specific demethylase 3A, lysine-specific demethylase 3A, testis-specific protein A, TSGADKFZp686P07111 | |
Mouse | |
IgG1 | |
Protein G purified | |
Chromatin Modifiers, Epigenetics | |
Primary | |
55818 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:250 | |
Purified | |
KDM3A | |
Purified recombinant fragment of human KDM3A (AA: 311-445) expressed in E. Coli. | |
105.4 kDa | |
0.1 ml | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title