Learn More
Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A Mouse anti-Human, HRP, Clone: 1000000000000, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP151678H
Catalog No. NBP151678H
Add to cart
Description
Description
Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A Monoclonal antibody specifically detects Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A
1000000000000
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
KDM3A
Purified recombinant fragment of human JMJD1A expressed in E. coli. [UniProt# Q9Y4C1]
Ammonium sulfate precipitation
RUO
Store at 4C in the dark.
Monoclonal
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
HRP
PBS with No Preservative
EC 1.14.11, EC 1.14.11.-, JHDM2AJMJD1AJHMD2A, JmjC domain-containing histone demethylation protein 2A, JMJD1, jumonji C domain-containing histone demethylase 2A, jumonji domain containing 1, jumonji domain containing 1A, Jumonji domain-containing protein 1A, KIAA0742DKFZp686A24246, lysine (K)-specific demethylase 3A, lysine-specific demethylase 3A, testis-specific protein A, TSGADKFZp686P07111
Mouse
IgG1
0.1 ml
Chromatin Modifiers, Epigenetics
Primary
55818