Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lysis Buffer, 0.5% Tween 20, pH 8.0

For isolation of His-tag proteins

Manufacturer:  Alfa Aesar™ J61792AP

Catalog No. AAJ61792AP


Order in Bulk

Request bulk or custom formats

Add to cart

Specifications

Specifications

500mL
Cell Lysis Reagent
8
Liquid
For isolation of His-tag modified proteins
Liquid
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit