Learn More
Aviva Systems Biology Corp LYSMD4 Antibody - N-terminal region (ARP52982_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52982_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LYSMD4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LYSMD4. Alternate names: FLJ33008, MGC99501ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60887