Learn More
Cayman ChemicalSupplier Diversity Partner Lyso-Sphingomyelin (d18:1), 1670-26-4, 1 mg DFS Item
Manufacturer:  Cayman ChemicalSupplier Diversity Partner 100079471
A naturally-occurring lipid which is produced by the enzymatic removal of the acylated group of sphingomyelin; serves as a substrate for autotaxin to produce sphingosine-1-phosphate; occurs naturally in plasma, is a constituent of lipoproteins, and is increased in some diseases
Catalog No. NC1472669