Learn More
Gibco™ Lysobacter Enzymogenes Arg-C Recombinant Protein, PeproTech®
SDP
Insect cells Recombinant Protein
Supplier:  Gibco™ 45054100UG
Encompass
 View more versions of this product
Catalog No. 45054100UG
Add to Cart
This item is not returnable. View return policy
Description
Description
Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides. Most proteases act in a specific manner, hydrolyzing bonds at, or adjacent to specific residues, or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes, including prenatal and postnatal development, reproduction, signal transduction, the immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, frequently used in the analysis and production of proteins. Arg-C specifically cleaves at the carboxyl side of Arginine residues. Arg-C has a sulfhydryl requirement; it is activated by dithiothreitol, cysteine, or other sulfhydryl-containing reagents. The presence of calcium ions is essential. The enzyme is inhibited by oxidizing agents and sulfhydryl reactants, and by Co^2+, Cu2+, Cd^2+, and heavy metal ions.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Functional Assay
26.8
5 x 20 μg
-20°C
<1 EU/μg
Unconjugated
GVGDIGSSDY CEKDIVCRVK PSAEFLSASK SVARMVFTPK TGYTGYCSGT LLNNSNSPKR QLFWSAAHCI STQKVANTLQ TYWLYDATGC DNDTLSDKAV TLTGGATLLH SHATRDTLLL ELKSAPPSGA YYAGWNSSAI ATKGTAIEGI HHPSGDLKKY SLGSVTALSS TIDGKKPLTK VAWTTGVTEG GSSGSGLFTI SSTSGYQLRG GLYGGTSYCS APSDPDYYSQ LDGVWSSIKT YFSPHHHHHH HH
≥ 98% by SDS-PAGE gel and HPLC analyses.
protein with no preservative
Lysobacter Enzymogenes Arg-C
Insect cells
RUO
Bacteria
Recombinant
Lyophilized
Recommendations
Recommendations