Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Lysobacter Enzymogenes Arg-C Recombinant Protein, PeproTech®
Insect cells Recombinant Protein
Supplier: Gibco™ 45054500UG
This item is not returnable.
View return policy
Description
Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides. Most proteases act in a specific manner, hydrolyzing bonds at, or adjacent to specific residues, or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes, including prenatal and postnatal development, reproduction, signal transduction, the immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, frequently used in the analysis and production of proteins. Arg-C specifically cleaves at the carboxyl side of Arginine residues. Arg-C has a sulfhydryl requirement; it is activated by dithiothreitol, cysteine, or other sulfhydryl-containing reagents. The presence of calcium ions is essential. The enzyme is inhibited by oxidizing agents and sulfhydryl reactants, and by Co^2+, Cu2+, Cd^2+, and heavy metal ions.Specifications
Functional Assay | |
26.8 | |
25 x 20 μg | |
-20°C | |
<1 EU/μg | |
Unconjugated | |
GVGDIGSSDY CEKDIVCRVK PSAEFLSASK SVARMVFTPK TGYTGYCSGT LLNNSNSPKR QLFWSAAHCI STQKVANTLQ TYWLYDATGC DNDTLSDKAV TLTGGATLLH SHATRDTLLL ELKSAPPSGA YYAGWNSSAI ATKGTAIEGI HHPSGDLKKY SLGSVTALSS TIDGKKPLTK VAWTTGVTEG GSSGSGLFTI SSTSGYQLRG GLYGGTSYCS APSDPDYYSQ LDGVWSSIKT YFSPHHHHHH HH | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
protein with no preservative | |
Lysobacter Enzymogenes Arg-C | |
Insect cells | |
RUO | |
Bacteria | |
Recombinant | |
Lyophilized |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction