Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysozyme, Chicken Egg White, Ultrapure, Affymetrix/USB™

Click to view available options
Quantity:
5g
10g
25g
100g
Packaging:
Plastic bottle
12650-88-3
C125H196N40O36S2
2899.307
MFCD00131557
JFXJPYIEDZSWNF-JWBGUOTLSA-N
lysozyme, chicken egg white,unii-1zml031y3c,lysozyme chicken egg white,unii-968jka7t33
131750181
CC(C)CC(C(=O)NC(CC(=O)O)C(=O)NC(CC(=O)N)C(=O)NC(CC1=CC=C(C=C1)O)C(=O)NC(CCCNC(=N)N)C(=O)NCC(=O)NC(CC2=CC=C(C=C2)O)C(=O)NC(CO)C(=O)NC(CC(C)C)C(=O)NCC(=O)NC(CC(=O)N)C(=O)O)NC(=O)CNC(=O)C(CC3=CN=CN3)NC(=O)C(CCCNC(=N)N)NC(=O)C(CCCCN)NC(=O)C(CCSC)NC(=O)C(C)NC(
12650-88-3
C125H196N40O36S2
2899.307
MFCD00131557
JFXJPYIEDZSWNF-JWBGUOTLSA-N
lysozyme, chicken egg white,unii-1zml031y3c,lysozyme chicken egg white,unii-968jka7t33
131750181
CC(C)CC(C(=O)NC(CC(=O)O)C(=O)NC(CC(=O)N)C(=O)NC(CC1=CC=C(C=C1)O)C(=O)NC(CCCNC(=N)N)C(=O)NCC(=O)NC(CC2=CC=C(C=C2)O)C(=O)NC(CO)C(=O)NC(CC(C)C)C(=O)NCC(=O)NC(CC(=O)N)C(=O)O)NC(=O)CNC(=O)C(CC3=CN=CN3)NC(=O)C(CCCNC(=N)N)NC(=O)C(CCCCN)NC(=O)C(CCSC)NC(=O)C(C)NC(
Quantity 25g
Packaging Plastic bottle
Physical Form Crystalline Powder
Grade Ultrapure
Chemical Name or Material Lysozyme