Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysozyme, Chicken Egg White, Ultrapure, Thermo Scientific™

Catalog Number p-7082845
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Plastic bottle
12650-88-3
C125H196N40O36S2
2899.307
MFCD00131557
JFXJPYIEDZSWNF-JWBGUOTLSA-N
lysozyme, chicken egg white, unii-1zml031y3c, lysozyme chicken egg white, unii-968jka7t33
131750181
CC(C)CC(C(=O)NC(CC(=O)O)C(=O)NC(CC(=O)N)C(=O)NC(CC1=CC=C(C=C1)O)C(=O)NC(CCCNC(=N)N)C(=O)NCC(=O)NC(CC2=CC=C(C=C2)O)C(=O)NC(CO)C(=O)NC(CC(C)C)C(=O)NCC(=O)NC(CC(=O)N)C(=O)O)NC(=O)CNC(=O)C(CC3=CN=CN3)NC(=O)C(CCCNC(=N)N)NC(=O)C(CCCCN)NC(=O)C(CCSC)NC(=O)C(C)NC(
12650-88-3
C125H196N40O36S2
2899.307
MFCD00131557
JFXJPYIEDZSWNF-JWBGUOTLSA-N
lysozyme, chicken egg white, unii-1zml031y3c, lysozyme chicken egg white, unii-968jka7t33
131750181
CC(C)CC(C(=O)NC(CC(=O)O)C(=O)NC(CC(=O)N)C(=O)NC(CC1=CC=C(C=C1)O)C(=O)NC(CCCNC(=N)N)C(=O)NCC(=O)NC(CC2=CC=C(C=C2)O)C(=O)NC(CO)C(=O)NC(CC(C)C)C(=O)NCC(=O)NC(CC(=O)N)C(=O)O)NC(=O)CNC(=O)C(CC3=CN=CN3)NC(=O)C(CCCNC(=N)N)NC(=O)C(CCCCN)NC(=O)C(CCSC)NC(=O)C(C)NC(

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Packaging Plastic bottle
Quantity 10 g
Grade Ultrapure
Physical Form Crystalline Powder
Chemical Name or Material Lysozyme
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit