Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysozyme Rabbit anti-Human, Mouse, Allophycocyanin/Cy7, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP261118APCCY7

Catalog No. N261118APC7


Add to cart

Description

Description

Lysozyme Polyclonal antibody specifically detects Lysozyme in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

Lysozyme
APC-Cyanine7
PBS with 0.05% Sodium Azide
EC 3.2.1.17, lysozyme, lysozyme (renal amyloidosis)1,4-beta-N-acetylmuramidase C, lysozyme C, LZM, renal amyloidosis
Rabbit
IgG
0.1 mL
Store at 4°C in the dark. Do not freeze.
Polyclonal
Human, Mouse
Flow Cytometry
Flow Cytometry
Affinity Purified, Conjugated
LYZ
Full length recombinant human Lysozyme protein.
Immunogen affinity purified
Cardiovascular Biology, Immunology
Primary
4069.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit