Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LYST Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565778
Description
Immunogen sequence: KKYETEEGVNK AAWQKTVNNN QQSLFQRLDS KSKDISKIAA DITQAVSLSQ GNERKKVIQH IRGMYKVDLS ASRHWQELIQ QLTHDRAVWY DPIYYPTSWQ Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.
May be required for sorting endosomal resident proteins into late multivesicular endosomes by a mechanism involving microtubules.
Specifications
LYST | |
Polyclonal | |
Unconjugated | |
LYST | |
Beige homolog; Chediak-Higashi syndrome 1; CHS; CHS1; lysosomal trafficking regulator; lysosomal-trafficking regulator; LYST | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1130 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99698 | |
LYST | |
Recombinant Human LYST. Recombinant protein control fragment (Product #RP-106457). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction