Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYZL1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP286704

Catalog No. NB034326


Add to cart

Description

Description

LYZL1 Polyclonal specifically detects LYZL1 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

LYZL1
Unconjugated
PBS, 2% Sucrose
EC 3.2.1.17, KAAG648, LYC2bA534G20.1, lysozyme-like 1, lysozyme-like protein 1, MGC33408, PRO1278
The immunogen is a synthetic peptide directed towards the middle region of mouse LYZL1
100 μg
Lipid and Metabolism
Primary
84569
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Immunogen affinity purified
RUO
Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles.
Polyclonal
Mouse
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit