Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LZIC Mouse anti-Human, Clone: OTI3A7, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526212

Catalog No. PIMA526212


Add to cart

Description

Description

LZIC belongs to the CTNNBIP1 family. It is ubiquitously expressed with highest levels in kidney. LZIC is up-regulated in several cases of gastric cancers.
Specifications

Specifications

LZIC
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8WZA0
Protein LZIC; Leucine zipper and CTNNBIP1 domain-containing protein; Leucine zipper and ICAT homologous domain-containing protein; LZIC
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84328
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI3A7
Unconjugated
LZIC
Liquid
LZIC
Human recombit protein fragment corresponding to amino acids 1-190 of LZIC produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit