Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LZK Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549765
Description
The antibody detects endogenous levels of total LZK / MAP3K13 protein.
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The tyrosine-like kinase (TLK) group consists of 40 tyrosine and serine-threonine kinases such as MLK (mixed-lineage kinase), LISK (LIMK/TESK), IRAK (interleukin-1 receptor-associated kinase), Raf, RIPK (receptor-interacting protein kinase), and STRK (activin and TGF-beta receptors) families.
Specifications
LZK | |
Polyclonal | |
Unconjugated | |
MAP3K13 | |
C130026N12Rik; Leucine zipper-bearing kinase; LZK; Map3k13; MEKK13; mitogen activated protein kinase kinase kinase 13; mitogen-activated protein kinase kinase kinase 13; Mixed lineage kinase; MLK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
303823, 71751, 9175 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O43283, Q1HKZ5 | |
MAP3K13 | |
Synthesized peptide derived from internal of human M3K13. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction