Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OIT3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551204

Catalog No. PA551204


Add to cart

Description

Description

The antibody detects endogenous levels of total LZP / OIT3 protein.

May be involved in hepatocellular function and development.
Specifications

Specifications

OIT3
Polyclonal
Unconjugated
OIT3
Liquid
OIT3
Fusion protein of human OIT3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.4 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q8WWZ8
LZP
Rabbit
IgG
100 μL
-20°C
Primary
170392
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit