Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OIT3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548203

Catalog No. PA548203

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be involved in hepatocellular function and development.
Specifications

Specifications

OIT3
Polyclonal
PBS with 0.09% sodium azide
Q8WWZ8, Q8R4V5
Oncoprotein-induced transcript 3 protein, Liver-specific zona pellucida domain-containing protein, OIT3, LZP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
170392, 18302
Western Blot
Unconjugated
OIT3
Liquid
OIT3
Gst fusion protein from human OIT3.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit