Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ M 084 hydrochloride
TRPC4/5 channel blocker; antidepressant and anxiolytic
$119.48 - $498.80
Specifications
Chemical Name or Material | N-Butyl-1H-benzimidazol-2-amine hydrochloride |
---|---|
CAS | 1992047-63-8 |
Target | TRPC Channel Blockers |
Molecular Formula | C11H15N3.HCl |
Formula Weight | 225.72 |
Description
TRPC4/5 channel blocker (IC50 values are 3.7-10.3 and 8.2 μM, respectively). Also weakly blocks TRPC3 channels. Exhibits rapid antidepressant and anxiolytic effects in vivo.Specifications
N-Butyl-1H-benzimidazol-2-amine hydrochloride | |
TRPC Channel Blockers | |
225.72 |
1992047-63-8 | |
C11H15N3.HCl | |
>99% |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title